Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8/18/19 Antibody (2A4), Novus Biologicals™
SDP

Catalog No. NB036544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036544 Supplier Novus Biologicals Supplier No. NBP288953
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 8/18/19 Monoclonal specifically detects Cytokeratin 8/18/19 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cytokeratin 8/18/19
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 2A4
Conjugate Unconjugated
Formulation PBS containing 1% BSA
Gene Alias 40-kDa keratin intermediate filament, CARD2, Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CK19, CK-19, CK8, CK-8, CYK18, CYK8, cytokeratin 18, Cytokeratin-18, Cytokeratin-8, K18, K19, K1CS, K2C8, K8, Keratin 18, keratin 19, keratin 8, keratin, type I cytoskeletal 18, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin, type II cytoskeletal 8, Keratin-18, Keratin-19, keratin-8, KO, MGC15366, Type-II keratin Kb8
Host Species Mouse
Immunogen Cytokeratins isolated from cultured porcine kidney epithelial cells (LLC-PK).
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3856
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.