Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8/18 Antibody (5D3), Novus Biologicals™
SDP

Catalog No. NB12017139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12017139 1 mL
NB018083 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB12017139 Supplier Novus Biologicals Supplier No. NB12017139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 8/18
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5D3
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Gene Alias CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8
Gene Symbols KRT8
Host Species Mouse
Immunogen Cytokeratins from the human breast carcinoma cell line MCF-7.
Purification Method Tissue culture supernatant
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3856
Test Specificity This antibody is specific to human cytokeratins 8 (52.5 kDa) and 18 (45 kDa). It stains simple and glandular epithelium. It can be useful for the identification of adenocarcinomas and most squamous cell carcinomas.
Target Species Human
Content And Storage Store at 4°C.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.