Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Antibody (5D3), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$268.00 - $816.50
Specifications
Antigen | Cytokeratin 8/18 |
---|---|
Clone | 5D3 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018083
![]() |
Novus Biologicals
NB120171390.25ML |
0.25 mL |
Each for $268.00
|
|
|||||
NB12017139
![]() |
Novus Biologicals
NB12017139 |
1 mL |
Each for $816.50
|
|
|||||
Description
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 8/18 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cytoskeleton Markers | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
KRT8 | |
IgG1 | |
Tissue culture supernatant | |
This antibody is specific to human cytokeratins 8 (52.5 kDa) and 18 (45 kDa). It stains simple and glandular epithelium. It can be useful for the identification of adenocarcinomas and most squamous cell carcinomas. |
5D3 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
3856 | |
Cytokeratins from the human breast carcinoma cell line MCF-7. | |
Primary | |
Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title