Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8 Antibody (OTI1B12), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289105 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289105 Supplier Novus Biologicals Supplier No. NBP271082APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 8 Monoclonal antibody specifically detects Cytokeratin 8 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Cytokeratin 8
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI1B12
Conjugate APC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias CARD2, CK8, CK-8, CYK8, Cytokeratin-8, K2C8, K8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 91-381 of human CK8 (NP_002264) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3856
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.