| Antigen |
Cytokeratin 8 |
| Applications |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification |
Monoclonal |
| Clone |
TS1 |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml, SDS-Page |
| Formulation |
PBS with 0.05% BSA. with 0.05% Sodium Azide |
| Gene Accession No. |
P05787 |
| Gene Alias |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 |
| Gene Symbols |
KRT8 |
| Host Species |
Mouse |
| Immunogen |
Keratin preparation from a human carcinoma |
| Molecular Weight of Antigen |
52.5 kDa |
| Purification Method |
Protein G purified |
| Quantity |
0.02 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Cancer, Cytoskeleton Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
3856 |
| Test Specificity |
Epitope of this MAb is located between aa343-357 (ELAIKDANAKLSELE). Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). CK8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin 8 and 18. CK8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine, trachea, and esophagus as well as in transitional epithelium. Anti-CK8 does not react with skeletal muscle or nerve cells. Epithelioid sarcoma, chordoma, and adamantinoma show strong positivity corresponding to that of simple epithelia (with antibodies against CK8, CK18 and CK19). Reportedly, anti-CK8 is useful for the differentiation of lobular (ring-like, perinuclear) from ductal (peripheral-predominant) carcinoma of the breast. |
| Target Species |
Human, Ferret (Negative), Rat (Negative) |
| Content And Storage |
Store at 4C. |
| Form |
Purified |
| Isotype |
IgG1 κ |
|
Show More
Show Less
|
|