Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, Acidic (Type I or LMW) Antibody (AE1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB002749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002749 Supplier Novus Biologicals Supplier No. NBP262221AF532

Mouse Monoclonal Antibody

Cytokeratin, Acidic (Type I or LMW) Monoclonal specifically detects Cytokeratin, Acidic (Type I or LMW) in Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.

Specifications

Antigen Cytokeratin, Acidic (Type I or LMW)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone AE1
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CK10, CK14, CK15, CK16, CK19, cytokeratin 10, cytokeratin 14, cytokeratin 15, cytokeratin 16, cytokeratin 19
Host Species Mouse
Immunogen Human epidermal keratins
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity The antibody MEM-181 reacts with CD25 (Interleukin-2 receptor alpha chain), a 55 kDa type I transmembrane glycoprotein expressed on activated B and T lymphocytes, activated monocytes/macrophages and on CD4+ T lymphocytes (T regulatory cells); it is lost on resting B and T lymphocytes.
Target Species Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.