Learn More
Cytokeratin, Basic (Type II or HMW) Antibody (SPM116), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytokeratin, Basic (Type II or HMW) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM116 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | CK1, CK10, CK14, CK5, cytokeratin 1, cytokeratin 10, cytokeratin 14, cytokeratin 5 |
| Host Species | Mouse |
| Immunogen | Human epidermal keratin (Uniprot: Q01546) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.