Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, HMW Antibody (KRTH/1576R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP25316420UG
Description
Cytokeratin, HMW Monoclonal specifically detects Cytokeratin, HMW in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin, HMW | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml | |
CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
Rabbit | |
60 kDa | |
20 μg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
KRTH/1576R | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
KRT76 | |
Recombinant full-length human KRT76 protein | |
Protein A or G purified | |
RUO | |
51350 | |
Store at 4C. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction