Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, HMW Antibody (KRTH/1576R), Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50 - $548.00
Specifications
Antigen | Cytokeratin, HMW |
---|---|
Clone | KRTH/1576R |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25316420
![]() |
Novus Biologicals
NBP25316420UG |
20 μg |
Each for $304.50
|
|
|||||
NB253164100
![]() |
Novus Biologicals
NBP253164100UG |
100 μg |
Each for $548.00
|
|
|||||
NB253164200
![]() |
Novus Biologicals
NBP253164200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Cytokeratin, HMW Monoclonal specifically detects Cytokeratin, HMW in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin, HMW | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
KRT76 | |
IgG κ | |
Protein A or G purified | |
60 kDa |
KRTH/1576R | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
51350 | |
Recombinant full-length human KRT76 protein | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title