Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, LMW Antibody (rKRTL/6616), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB391491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391491 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391491 Supplier Novus Biologicals Supplier No. NBP308674C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin, LMW Monoclonal antibody specifically detects Cytokeratin, LMW in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin, LMW
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rKRTL/6616
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39
Host Species Mouse
Immunogen Synthetic peptide corresponding to Cytokeratin, LMW residues within aa100-200 of Cytokeratin, LMW (Uniprot: Q7Z794)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374454
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.