Learn More
Description
Specifications
Specifications
| Antigen | pan Cytokeratin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | C-11 |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 594 |
| Dilution | Western Blot 1:1000, Flow Cytometry 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Flow (Intracellular) 1 ug/mL, CyTOF-ready |
| Formulation | PBS (pH 7.4) with 15mM Sodium Azide |
| Gene Alias | CK1, EHK, EHK1, EPPK, K1, Keratin 1, KRT1, KRT1A, NEPPK |
| Gene Symbols | KRT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
