Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, pan Antibody (KRT/1877R), PerCP, Novus Biologicals™
SDP

Catalog No. NB387266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387266 Supplier Novus Biologicals Supplier No. NBP308398PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Guinea Pig, Goat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin, pan
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT/1877R
Conjugate PerCP
Formulation PBS
Gene Alias AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK
Host Species Rabbit
Immunogen Keratin-enriched preparation from cultured human epithelial cells
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Target Species Human, Mouse, Rat, Pig, Amphibian
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.