Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, pan Antibody, Novus Biologicals™
SDP

Catalog No. NB600579 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600579 0.5 mL
NB600579SS 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600579 Supplier Novus Biologicals Supplier No. NB600579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

Cytokeratin, pan Polyclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Bovine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen pan Cytokeratin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:75 - 1:200
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Symbols KRT1
Host Species Rabbit
Immunogen This Cytokeratin, pan Antibody was developed against pan Cytokeratin isolated from bovine muzzle epidermis.
Purification Method Protein A purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Test Specificity NB600-579 has cross-reactivity with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa molecular weight. The antibody is well suited for the staining of a broad spectrum of human keratins.
Target Species Human, Mouse, Bovine, Monkey
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.