Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, pan Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$207.00 - $519.67
Specifications
| Antigen | pan Cytokeratin |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:75 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600579SS
![]() |
Novus Biologicals
NB600579SS |
0.1 mL |
Each for $207.00
|
|
|||||
NB600579
![]() |
Novus Biologicals
NB600579 |
0.5 mL |
Each for $519.67
|
|
|||||
Description
Cytokeratin, pan Polyclonal specifically detects Cytokeratin, pan in Human, Mouse, Bovine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| pan Cytokeratin | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| KRT1 | |
| IgG | |
| Protein A purified | |
| NB600-579 has cross-reactivity with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa molecular weight. The antibody is well suited for the staining of a broad spectrum of human keratins. |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:75 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Bovine, Monkey | |
| 3848 | |
| This Cytokeratin, pan Antibody was developed against pan Cytokeratin isolated from bovine muzzle epidermis. | |
| Primary | |
| Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title