Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, pan Antibody (PCK-26), Novus Biologicals™
SDP

Catalog No. NB1206401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206401 Supplier Novus Biologicals Supplier No. NB1206401
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Fish, Guinea Pig, Goat, Hamster, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen pan Cytokeratin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCK-26
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300, Immunohistochemistry-Paraffin 1:300 - 1:600, Immunohistochemistry-Frozen 1:300 - 1:600
Gene Accession No. P04264
Gene Symbols KRT1
Host Species Mouse
Immunogen This Cytokeratin, pan Antibody (PCK-26) was developed against the full length native protein (purified) (Human epidermis).
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Test Specificity PCK-26 recognizes the 58 kDa cytokeratin 5, the 56 kDa cytokeratin 6 and the 52 kDa cytokeratin 8 band in immunoblotting (Lane EB et al). The antibody recognizes an epitope located on the Type II cytokeratins 1, 5, 6, and 8. PCK-26 is a broad spectrum antibody which reacts specifically with a variety of normal, reactive, and neoplastic epithelial tissues. The antibody reacts with simple, cornifying, and non-cornifying squamous epithelia and pseudostratified epithelia.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Goat, Hamster, Rabbit, Reptile, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.