Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, pan Rabbit anti-Human, Clone: Cocktail PCK/4933R, Novus Biologicals™
SDP

Catalog No. NB177777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177777 20 μg
NB177776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177777 Supplier Novus Biologicals Supplier No. NBP31391020UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin, pan
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Cocktail PCK/4933R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Host Species Rabbit
Immunogen Crude cytokeratin extract prepared from RT-4 and MCF-7 cells.
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.