Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus Antibody (HCMV16 (51C1)), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB164398AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164398AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164398AF Supplier Novus Biologicals Supplier No. NB10064398AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus (CMV), Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen Cytomegalovirus
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone HCMV16 (51C1)
Conjugate Alexa Fluor 405
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CMV; HHV 5; HHV5; Human herpesvirus 5
Host Species Mouse
Immunogen Cytomegalovirus extracts
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity NB100-64398 is specific for Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells. It does not react with HSV, VZV, EBV or uninfected cells.
Target Species Cytomegalovirus (CMV), Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.