Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus Antibody (HCMV16 (51C1)) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017574 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017574 0.025 mg
NB10064398 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017574 Supplier Novus Biologicals Supplier No. NB100643980.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus (CMV), Virus samples. It is validated for ELISA, Functional, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen Cytomegalovirus
Applications ELISA, Functional Assay, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone HCMV16 (51C1)
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with 0.09% Sodium Azide
Gene Alias CMV; HHV 5; HHV5; Human herpesvirus 5
Host Species Mouse
Immunogen Cytomegalovirus extracts
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64398 is specific for Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells. It does not react with HSV, VZV, EBV or uninfected cells.
Target Species Cytomegalovirus (CMV), Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.