Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus gB Antibody (hCMV34), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB10064532W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064532W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064532W Supplier Novus Biologicals Supplier No. NB10064532AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytomegalovirus gB Monoclonal specifically detects Cytomegalovirus gB in Cytomegalovirus (CMV), Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Cytomegalovirus gB
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone hCMV34
Conjugate Alexa Fluor 405
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CMV, CMV glycoprotein B, CMV glycoprotein GP55, Cytomegalovirus glycoprotein GP55, gB, Glycoprotein B, Glycoprotein GP55, HCMV, HCMV glycoprotein B, HCMV glycoprotein GP55, HHV 5 glycoprotein B, HHV 5 glycoprotein GP55, HHV5 glycoprotein B, HHV5 glycoprotein GP55, Human herpesvirus 5 glycoprotein B, Human herpesvirus 5 glycoprotein GP55, UL55
Host Species Mouse
Immunogen Native
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Recognizes cytomegalovirus glycoprotein B, a viral ligand for CD209 which allows capture of viral particles by dendritic cells. This does not react with HSV, VSV or EBV.
Target Species Cytomegalovirus (CMV), Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.