Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus gH Antibody (HCMV16), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB340450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340450 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340450 Supplier Novus Biologicals Supplier No. NBP320157MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytomegalovirus gH Monoclonal antibody specifically detects Cytomegalovirus gH in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Neutralization

Specifications

Antigen Cytomegalovirus gH
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone HCMV16
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Neutralization
Formulation 50mM Sodium Borate
Gene Alias envelope glycoprotein H, gH, gpUL75, HCMV gH, HHV-5 gH, HHV5 glycoprotein H, Human betaherpesvirus 5 envelope glycoprotein H, Human Cytomegalovirus glycoprotein H, UL75
Host Species Mouse
Immunogen The original antibody was generated by immunization of mice with purified HCMV strain AD169.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3077450
Target Species Cytomegalovirus (CMV), Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.