Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus p65 Antibody (CMV100), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP234595Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234595Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234595Z Supplier Novus Biologicals Supplier No. NBP234595AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytomegalovirus p65 Monoclonal specifically detects Cytomegalovirus p65 in Cytomegalovirus, Virus samples. It is validated for ELISA.

Specifications

Antigen Cytomegalovirus p65
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CMV100
Conjugate Alexa Fluor 700
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias CMV, p65
Host Species Mouse
Immunogen Recombinant CMV-p65 protein
Molecular Weight of Antigen 65 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.
Target Species Cytomegalovirus (CMV), Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.