Learn More
Description
Specifications
Specifications
| Antigen | Cytomegalovirus pp65 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | B051M |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Gene Alias | 64 kDa lower matrix phosphoprotein, 65 kDa lower matrix phosphoprotein, CMV, CMV 65 kDa lower matrix phosphoprotein, CMV pp65, CMV pp65 IE, CMV tegument protein UL83, cytomegalovirus, Cytomegalovirus 65 kDa lower matrix phosphoprotein, Cytomegalovirus pp65, Cytomegalovirus tegument protein UL83, GP64, PP64, PP65, Tegument protein UL 83, Tegument protein UL83, UL 83, UL83 |
| Host Species | Mouse |
| Immunogen | CMV Lysate |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
