Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosine, 99+%

Catalog No. ac161760050
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00006034
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
PubChem CID 597
ChEBI CHEBI:16040
SMILES NC1=CC=NC(=O)N1

Specifications

Melting Point >300.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24,314
Merck Index 15,2792
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkalines and acids,slightly soluble in ethanol,chloroform,,insoluble in ether and hydrocarbons
Spectrophotometry 98% min. (lambda max. 275nm,pH 1) molar absorptivity 10400+/-2%,on dry basis
IUPAC Name 6-amino-1H-pyrimidin-2-one
Formula Weight 111.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytosine
Show More Show Less
EINECSNumber 200-749-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.