Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody (15E5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222557 Supplier Novus Biologicals Supplier No. NBP222557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Monoclonal specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1500000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 2-5ug, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500
Gene Accession No. NP_060705
Gene Alias CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1
Gene Symbols CNDP2
Host Species Mouse
Immunogen Recombinant human CNDP2 (1-475aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 55748
Test Specificity Anti-human CNDP2 mAb, clone 15E5, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CNDP2 protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.