Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody (210), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB039658 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB039658 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB039658 Supplier Novus Biologicals Supplier No. NBP290495R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Monoclonal specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 210
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL (NP_075638.2; Met1-Asn475).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 55748
Test Specificity Has cross-reactivity in ELISA with: Human CNDP2
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.