Learn More
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody (OTI8A2), PerCP, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI8A2 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1 |
| Gene Symbols | CNDP2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.