Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic beta-Glucosidase/GBA3 Antibody (008), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB110106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB110106 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB110106 Supplier Novus Biologicals Supplier No. NBP289924AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cytosolic beta-Glucosidase/GBA3
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 8
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met1-Leu469).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57733
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.