Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic beta-Glucosidase/GBA3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB089753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089753 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089753 Supplier Novus Biologicals Supplier No. NBP299838R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytosolic beta-Glucosidase/GBA3 Polyclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for ELISA.

Specifications

Antigen Cytosolic beta-Glucosidase/GBA3
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met 1-Leu 469).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57733
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.