Learn More
Cytosolic Sulfotransferase 1B1/SULT1B1 Antibody, DyLight 650, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytosolic Sulfotransferase 1B1/SULT1B1 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B2, Thyroid hormone sulfotransferase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 1B1/SULT1B1 (NP_055280.2; Leu 2-Ile 296). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.