Learn More
Description
Specifications
Specifications
| Antigen | Cytosolic Sulfotransferase 1B1/SULT1B1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | O43704 |
| Gene Alias | EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B2, Thyroid hormone sulfotransferase |
| Gene Symbols | SULT1B1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
