Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 1B1/SULT1B1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Cytosolic Sulfotransferase 1B1/SULT1B1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154403
SDP
View Documents
Novus Biologicals
NBP154403
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Cytosolic Sulfotransferase 1B1/SULT1B1 Polyclonal specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Cytosolic Sulfotransferase 1B1/SULT1B1
Polyclonal
Rabbit
O43704
27284
Synthetic peptides corresponding to SULT1B1(sulfotransferase family, cytosolic, 1B, member 1) The peptide sequence was selected from the N terminal of SULT1B1. Peptide sequence KRGFITEKVPMLEMTLPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFW.
Primary
Western Blot
Unconjugated
RUO
EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B2, Thyroid hormone sulfotransferase
SULT1B1
IgG
35 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.