Learn More
Cytosolic Sulfotransferase 1C2/SULT1C2 Antibody (OTI4G1), Janelia Fluor™ 549, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytosolic Sulfotransferase 1C2/SULT1C2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI4G1 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1CNo.1 |
| Gene Symbols | SULT1C2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.