Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB103154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB103154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB103154 Supplier Novus Biologicals Supplier No. NBP297722AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytosolic Sulfotransferase 2A1/SULT2A1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Cytosolic Sulfotransferase 2A1/SULT2A1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.