Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 2A1/SULT2A1 Rabbit anti-Human, Clone: 4, Novus Biologicals™
SDP

Catalog No. NB021258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021258 100 μg
NB021259 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021258 Supplier Novus Biologicals Supplier No. NBP289896100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytosolic Sulfotransferase 2A1/SULT2A1 Monoclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for ELISA.

Specifications

Antigen Cytosolic Sulfotransferase 2A1/SULT2A1
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1
Host Species Rabbit
Immunogen Recombinant Human Cytosolic Sulfotransferase 2A1/SULT2A1 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.