Learn More
Cytosolic Sulfotransferase 2B1/SULT2B1 Antibody (007), DyLight 550, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytosolic Sulfotransferase 2B1/SULT2B1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 7 |
| Conjugate | DyLight 550 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 2B1/SULT2B1 (NP_004596.2; Asp 2-Ser 365) . |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.