Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 2B1/SULT2B1 Antibody (007), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB335113 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335113 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335113 Supplier Novus Biologicals Supplier No. NBP289895MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytosolic Sulfotransferase 2B1/SULT2B1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Cytosolic Sulfotransferase 2B1/SULT2B1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 007
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 2B1/SULT2B1 (Accession#: NP_004596.2, Asp 2-Ser 365)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6820
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.