Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 2B1/SULT2B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232596 0.1 mL
NB436985 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP232596 Supplier Novus Biologicals Supplier No. NBP232596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytosolic Sulfotransferase 2B1/SULT2B1 Polyclonal specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytosolic Sulfotransferase 2B1/SULT2B1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1
Gene Symbols SULT2B1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: HIKGWLRMKGKDNFLFITYEELQQDLQGSVERICGFLGRPLGKEALGSVVAHSTFSAMKANTMSNYTLLPPSLLDHRR
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6820
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.