Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYYR1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP168916
Description
CYYR1 Polyclonal specifically detects CYYR1 in Rat samples. It is validated for Western Blot.Specifications
CYYR1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q5PQS5 | |
CYYR1 | |
Synthetic peptides corresponding to Cyyr1 (cysteine/tyrosine-rich 1) The peptide sequence was selected from the middle region of Cyyr1. Peptide sequence FVYAEDCRAQCGKDCRAYCCNGSTPHCCSYYAYIGSILSGTAIAGIVFGI. | |
Affinity Purified | |
RUO | |
116159 | |
Human, Mouse, Rat, Canine, Equine, Rabbit, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
C21orf95, cysteine and tyrosine-rich 1, cysteine and tyrosine-rich protein 1, cysteine/tyrosine-rich 1, Proline-rich domain-containing protein | |
Rabbit | |
18 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction