Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYYR1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | CYYR1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CYYR1 Polyclonal specifically detects CYYR1 in Rat samples. It is validated for Western Blot.Specifications
CYYR1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
C21orf95, cysteine and tyrosine-rich 1, cysteine and tyrosine-rich protein 1, cysteine/tyrosine-rich 1, Proline-rich domain-containing protein | |
CYYR1 | |
IgG | |
Affinity Purified | |
18 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q5PQS5 | |
116159 | |
Synthetic peptides corresponding to Cyyr1 (cysteine/tyrosine-rich 1) The peptide sequence was selected from the middle region of Cyyr1. Peptide sequence FVYAEDCRAQCGKDCRAYCCNGSTPHCCSYYAYIGSILSGTAIAGIVFGI. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title