Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D 4476
GREENER_CHOICE

Catalog No. 290210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-0210 10 mg
29-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-0210 Supplier Tocris Bioscience™ Supplier No. 2902/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective CK1 inhibitor. Also inhibits TGF-βRI

Selective inhibitor of casein kinase 1 (CK1) and TGF-β type-I receptor (ALK5) that displays > 20-fold selectivity over SAPK2/p38 and a much greater selectivity over all other protein kinases tested. Suppresses site-specific phosphorylation and nuclear exclusion of FOXO1a.

Chemical Identifiers

CAS 301836-43-1
Molecular Formula C23H18N4O3

Specifications

Quantity 10 mg
Formula Weight 398.41
Purity >99%
Molecular Weight (g/mol) 398.422
Chemical Name or Material D 4476
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.