Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ D-AP4
Click to view available options
Quantity:
10 mg
50 mg
Description
D-AP4 is a broad spectrum excitatory amino acid receptor antagonist. Also agonist at the quisqualate-sensitized AP6 site in hippocampus where it is less potent than L-AP4.
DL Mixture and L-isomer also available.
Specifications
Specifications
Chemical Name or Material | D-(-)-2-Amino-4-phosphonobutyric acid |
CAS | 78739-01-2 |
Assay Percent Range | >98% |
Quantity | 50 mg |
Solubility | Soluble to 100 mM in 1eq. NaOH |
Molecular Formula | C4H10NO5P |
Formula Weight | Observed MW: 183.1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction