Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Green No. 5, Spectrum™ Chemical
SDP

Catalog No. 18601968 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle

D1119, 4403-90-1, C28H20N2O8S2Na2

Spectrum™ Chemical D and C Green No. 5

Chemical Identifiers

CAS 4403-90-1
Molecular Formula C28H20N2Na2O8S2
Molecular Weight (g/mol) 622.57
InChI Key FPAYXBWMYIMERV-UHFFFAOYSA-L
IUPAC Name disodium 5-methyl-2-({4-[(4-methyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate
SMILES [Na+].[Na+].CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Color Blue/Green
Quantity 100 g
Chemical Name or Material D and C Green No. 5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.