Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Yellow No. 10, Spectrum™ Chemical
SDP

Catalog No. 18601992 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle

Catalog No. 18-601-992

Supplier: Spectrum Chemical Manufacturing Corporation DC2005KGBL

Only null left
Add to Cart
Add to Cart

DC200, 8004-92-0, C18H9NO8S2Na

Spectrum™ Chemical D and C Yellow No. 10

Chemical Identifiers

CAS 8004-92-0
Molecular Formula C18H9NNa2O8S2
Molecular Weight (g/mol) 477.37
MDL Number MFCD00080727
InChI Key FZUOVNMHEAPVBW-UHFFFAOYSA-L
IUPAC Name disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Specifications

CAS Max % 100%
Percent Purity 85%
Grade Reagent
Packaging Amber Glass Bottle
Color Yellow
Quantity 5 kg
Chemical Name or Material D and C Yellow No. 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.