Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Dimer Antibody (DD1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB359682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359682 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359682 Supplier Novus Biologicals Supplier No. NB1108374AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

D-Dimer Monoclonal antibody specifically detects D-Dimer in Human, Monkey, Baboon samples. It is validated for Western Blot, ELISA, Immunoassay

Specifications

Antigen D-Dimer
Applications Western Blot, ELISA, Immunoassay
Classification Monoclonal
Clone DD1
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunoassay
Formulation 50mM Sodium Borate
Gene Alias D Dimer
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Monkey, Baboon
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.