Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Dimer Antibody (DD2), Novus Biologicals™
SDP

Catalog No. NB1108376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1108376 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1108376 Supplier Novus Biologicals Supplier No. NB1108376
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

D-Dimer Monoclonal specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA, Immunoassay, Sandwich ELISA.

Specifications

Antigen D-Dimer
Applications Western Blot, ELISA, Immunoassay
Classification Monoclonal
Clone DD2
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA, Immunoassay, Sandwich ELISA 1:100-1:2000
Gene Alias D Dimer
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Test Specificity this is more specific to high molecular weight fibrin degradation products than to D-dimer. No cross-reaction with D-monomer.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.