Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Dimer Antibody (DD44), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB10073039L Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073039L 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073039L Supplier Novus Biologicals Supplier No. NB10073039AF488

Mouse Monoclonal Antibody

D-Dimer Monoclonal specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen D-Dimer
Applications Western Blot, ELISA
Classification Monoclonal
Clone DD44
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias D Dimer
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity D-dimer and high molecular weight fibrin degradation products, no cross-reactivity with fibrinogen
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.