Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Galacturonic acid monohydrate, 97%

Catalog No. AAJ6628214
Click to view available options
Quantity:
5 g
25 g

CAS: 91510-62-2 | C6H9O7

D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.

Solubility
Soluble in water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91510-62-2
Molecular Formula C6H9O7
Molecular Weight (g/mol) 0.00
MDL Number MFCD00071585
InChI Key AEMOLEFTQBMNLQ-DTEWXJGMSA-M
Synonym d-+-galacturonic acid monohydrate
PubChem CID 91872458
IUPAC Name (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid;hydrate
SMILES *

Specifications

Color White
Quantity 25 g
Beilstein 1727087
Merck Index 14,4337
Solubility Information Soluble in water
Optical Rotation +53° (c=10 in Water)
Formula Weight 212.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material D-Galacturonic acid monohydrate
EINECSNumber 211-682-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.