Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Glucose, ACS Reagent, anhydrous
GSA_VA

Catalog No. ac410955000
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
MDL Number MFCD00063684
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.0°C
Color White
Ignition Residue 0.02% max.
Loss on Drying 0.2% max. (105°C, 6 hrs)
Infrared Spectrum Authentic
Packaging Plastic Bottle
Molecular Formula C6H12O6
Quantity 500 g
Beilstein 31, 83
Merck Index 15, 4495
Specific Rotation Condition + 52.75 (25.00°C c=10,H2O,NH4OH)
Specific Rotation + 52.75
Solubility Information 0.005% max. Insoluble Matter
Molecular Weight (g/mol) 180.156
Formula Weight 180.16
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Glucose, Anhydrous, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.