Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucuronic acid, 98+%

Catalog No. p-7046445
Click to view available options
Quantity:
5 g
25 g

CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol

D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00077778
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
Synonym 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
PubChem CID 65041
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O

Specifications

Melting Point ∽158°C (decomposition)
Quantity 5 g
Beilstein 1727083
Merck Index 14,4465
Solubility Information Soluble in water.
Optical Rotation +35° (c=6 in Water)
Formula Weight 194.14
Percent Purity ≥98%
Chemical Name or Material D-Glucuronic acid
EINECSNumber 229-486-4
RTECSNumber LZ8836600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.