Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D(-)-Isoascorbic acid, 98%

Catalog No. p-150207
Click to view available options
Quantity:
500 g
2.5 kg

CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00005378
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
PubChem CID 54675810
ChEBI CHEBI:51438
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 167.0°C to 172.0°C
Color White to Yellow
Loss on Drying 0.4% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 5171
Specific Rotation Condition −17.25°(25°C c=10,H2O)
Specific Rotation − 17.25
Solubility Information Solubility in water: 100g/l. Other solubilities: soluble in alcohol,pyridine,moderately soluble in acetone,slightly soluble in glycerol
Formula Weight 176.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Isoascorbic acid, 98%
Show More Show Less
EINECSNumber 201-928-0
RTECSNumber KF3015000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.